Sodium hydroxide solution 5 mol/L EMPROVE EXPERT 1 * 1 L
Catalog # 1.37041.1002
Supplier: Merck
Frequently Bought Together
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Matrix™ Reagent Reservoirs
Supplier: THERMO MATRIX TECHNOLOGIES
VWR® Facial Tissues
Supplier: VWR Collection
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium sulphite 95.0-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
VWR®, Bottles, Narrow Neck, Round, LDPE, with Screw Caps
Supplier: VWR Collection
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Optical dissolved oxygen (DO) measurement with IQ Sensor Net
Supplier: WTW ONLINE