ContactUsLinkComponent
The biotinylated CD38 protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-CD38 antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surf 1 * 2 mg
Artikelnr. ACRBMBC-K010-2MG
Supplier:  ACRO Biosystems
The biotinylated CD38 protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-CD38 antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surf 1 * 2 mg
Artikelnr. ACRBMBC-K010-2MG
Supplier:  ACRO Biosystems
Frequently Bought Together