ContactUsLinkComponent
Human IL-31, His Tag IL1-H5247 is expressed from human 293 cells HEK293. It contains AA Ser 24 - Thr 164 Accession Q6EBC2-1. 1 * 1 mg
Catalog # ACRBIL1-H5247-1MG
Supplier:  ACRO Biosystems
Human IL-31, His Tag IL1-H5247 is expressed from human 293 cells HEK293. It contains AA Ser 24 - Thr 164 Accession Q6EBC2-1. 1 * 1 mg
Catalog # ACRBIL1-H5247-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together