ContactUsLinkComponent
SARS-CoV-2 S protein RBD V367F, His Tag SPD-S52H4 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 541 Accessi on QHD43416.1 V367F. 1 * 1 mg
Catalog # ACRBSPD-S52H4-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 S protein RBD V367F, His Tag SPD-S52H4 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 541 Accessi on QHD43416.1 V367F. 1 * 1 mg
Catalog # ACRBSPD-S52H4-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together