ContactUsLinkComponent
Human CD3ECD3D Heterodimer Protein, His TagTag Free CDD-H52W1 is expressed from human 293 cells HEK293. It contains AA Asp 23 - Asp 126 CD3E Phe 22 - Ala 105 CD3D Accession P07766-1 CD3E P04234-1 CD3D. 1 * 50 µG
Catalog # ACRBCDD-H52W1-50UG
Supplier:  ACRO Biosystems
Human CD3ECD3D Heterodimer Protein, His TagTag Free CDD-H52W1 is expressed from human 293 cells HEK293. It contains AA Asp 23 - Asp 126 CD3E Phe 22 - Ala 105 CD3D Accession P07766-1 CD3E P04234-1 CD3D. 1 * 50 µG
Catalog # ACRBCDD-H52W1-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together