ContactUsLinkComponent
HCoV-NL63 S1 protein, His Tag SIN-V52H3 is expressed from human 293 cells HEK293. It contains AA Cys 19 - Val 717 Accession Q6Q1S2- 1. 1 * 1 mg
Catalog # ACRBSIN-V52H3-1MG
Supplier:  ACRO Biosystems
HCoV-NL63 S1 protein, His Tag SIN-V52H3 is expressed from human 293 cells HEK293. It contains AA Cys 19 - Val 717 Accession Q6Q1S2- 1. 1 * 1 mg
Catalog # ACRBSIN-V52H3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together