Biotinylated Human IL-22, His,Avitag™ IL2-H8247 is expressed from human 293 cells HEK293. It contains AA Ala 34 - Ile 179 Accession Q9GZX6-1. 1 * 200 µG
Catalog # ACRBIL2-H8247200UG
Supplier: ACRO Biosystems
Biotinylated Human IL-22, His,Avitag™ IL2-H8247 is expressed from human 293 cells HEK293. It contains AA Ala 34 - Ile 179 Accession Q9GZX6-1. 1 * 200 µG
Catalog # ACRBIL2-H8247200UG
Supplier: ACRO Biosystems
Frequently Bought Together
VWR®, Microcentrifuge Tubes and Screw Caps
Supplier: VWR Collection
Sampling scoops, LaboPlast®
Supplier: BURKLE
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Square Bottles, PET
Supplier: LAB3 OWN PRODUCTION
VWR®, Cryoboxes, Half Size, Cardboard
Supplier: VWR Collection
Eppendorf twin.tec® PCR Plates 96-well, 250 µl
Supplier: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
Storage plates, 1,2 ml square well, low profile
Supplier: THERMO MATRIX TECHNOLOGIES
VWR® Bottles, Narrow Neck, PET, Square, with Screw Cap
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
PCR plates, 96-well, fully skirted, low profile, Thermo-Fast®
Supplier: Thermo Fisher Scientific
Multiple well cell culture plates, Costar®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION