ContactUsLinkComponent
Cynomolgus IL-1 Rrp2, His Tag IL2-C52H5 is expressed from human 293 cells HEK293. It contains AA Asp 20 - Arg 335 Accession EHH6170 2.1. 1 * 100 µG
Catalog # ACRBIL2-C52H5100UG
Supplier:  ACRO Biosystems
Cynomolgus IL-1 Rrp2, His Tag IL2-C52H5 is expressed from human 293 cells HEK293. It contains AA Asp 20 - Arg 335 Accession EHH6170 2.1. 1 * 100 µG
Catalog # ACRBIL2-C52H5100UG
Supplier:  ACRO Biosystems
Frequently Bought Together