ContactUsLinkComponent
Biotinylated Mouse IL-6 R alpha, His,Avitag ILR-M82E9 is expressed from human 293 cells HEK293. It contains AA Leu 20 - Glu 357 Acce ssion P22272-1. 1 * 200 µG
  ACRBILR-M82E9200UG
Biotinylated Mouse IL-6 R alpha, His,Avitag ILR-M82E9 is expressed from human 293 cells HEK293. It contains AA Leu 20 - Glu 357 Acce ssion P22272-1. 1 * 200 µG
  ACRBILR-M82E9200UG
Frequently Bought Together