ContactUsLinkComponent
HCoV-229E Nucleocapsid, His Tag NUN-V5147 is expressed from E.coli cells. It contains AA Met 1 - Asn 389 Accession P15130-1. 1 * 100 µG
Catalog # ACRBNUN-V5147100UG
Supplier:  ACRO Biosystems
HCoV-229E Nucleocapsid, His Tag NUN-V5147 is expressed from E.coli cells. It contains AA Met 1 - Asn 389 Accession P15130-1. 1 * 100 µG
Catalog # ACRBNUN-V5147100UG
Supplier:  ACRO Biosystems
Frequently Bought Together