ContactUsLinkComponent
Human EpCAM, Mouse IgG2a Fc Tag EPM-H5255 is expressed from human 293 cells HEK293. It contains AA Gln 24 - Lys 265 Accession AAH1 4785.1. 1 * 1 mg
Catalog # ACRBEPM-H5255-1MG
Supplier:  ACRO Biosystems
Human EpCAM, Mouse IgG2a Fc Tag EPM-H5255 is expressed from human 293 cells HEK293. It contains AA Gln 24 - Lys 265 Accession AAH1 4785.1. 1 * 1 mg
Catalog # ACRBEPM-H5255-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together