ContactUsLinkComponent
Rhesus macaque Cynomolgus TROP-2, His Tag TR2-R52H3 is expressed from human 293 cells HEK293. It contains AA Gln 31 - Thr 274 Acces sion XP_001114599.1. 1 * 1 mg
Catalog # ACRBTR2-R52H3-1MG
Supplier:  ACRO Biosystems
Rhesus macaque Cynomolgus TROP-2, His Tag TR2-R52H3 is expressed from human 293 cells HEK293. It contains AA Gln 31 - Thr 274 Acces sion XP_001114599.1. 1 * 1 mg
Catalog # ACRBTR2-R52H3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together