ContactUsLinkComponent
Cynomolgus TREM2, His Tag TR2-C52H3 is expressed from human 293 cells HEK293. It contains AA His 19 - Ser 174 Accession A0A2K5UY39- 1. 1 * 100 µG
Catalog # ACRBTR2-C52H3100UG
Supplier:  ACRO Biosystems
Cynomolgus TREM2, His Tag TR2-C52H3 is expressed from human 293 cells HEK293. It contains AA His 19 - Ser 174 Accession A0A2K5UY39- 1. 1 * 100 µG
Catalog # ACRBTR2-C52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together