ContactUsLinkComponent
Cynomolgus DLL3, His Tag DL3-C52H3 is expressed from human 293 cells HEK293. It contains AA Ala 27 - Arg 490 Accession A0A2K5WSR4-1 . 1 * 100 µG
Catalog # ACRBDL3-C52H3100UG
Supplier:  ACRO Biosystems
Cynomolgus DLL3, His Tag DL3-C52H3 is expressed from human 293 cells HEK293. It contains AA Ala 27 - Arg 490 Accession A0A2K5WSR4-1 . 1 * 100 µG
Catalog # ACRBDL3-C52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together