ContactUsLinkComponent
Human VEGF-D, His Tag VED-H5228 is expressed from human 293 cells HEK293. It contains AA Phe 93 - Ser 201 Accession AAH27948. 1 * 100 µG
Catalog # ACRBVED-H5228100UG
Supplier:  ACRO Biosystems
Human VEGF-D, His Tag VED-H5228 is expressed from human 293 cells HEK293. It contains AA Phe 93 - Ser 201 Accession AAH27948. 1 * 100 µG
Catalog # ACRBVED-H5228100UG
Supplier:  ACRO Biosystems
Frequently Bought Together