ContactUsLinkComponent
Biotinylated Human IL-13 R alpha 2, His,Avitag IL2-H82E6 is expressed from human 293 cells HEK293. It contains AA Asp 27 - Arg 343 A ccession Q14627-1. 1 * 25 µG
Catalog # ACRBIL2-H82E6-25UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-13 R alpha 2, His,Avitag IL2-H82E6 is expressed from human 293 cells HEK293. It contains AA Asp 27 - Arg 343 A ccession Q14627-1. 1 * 25 µG
Catalog # ACRBIL2-H82E6-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together