ContactUsLinkComponent
Human CD16b NA2, SUMO,His Tag CDB-H5294 is expressed from human 293 cells HEK293. It contains AA Gly 17 - Ser 200 Accession O75015 -1. null
Catalog # ACRBCDB-H5294-1MG
Supplier:  ACRO Biosystems
Human CD16b NA2, SUMO,His Tag CDB-H5294 is expressed from human 293 cells HEK293. It contains AA Gly 17 - Ser 200 Accession O75015 -1. null
Catalog # ACRBCDB-H5294-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together