ContactUsLinkComponent
Mouse MARCO, His Tag MAR-M5245 is expressed from human 293 cells HEK293. It contains AA Gln 70 - Ser 518 Accession Q60754-1. 1 * 100 µG
Catalog # ACRBMAR-M5245100UG
Supplier:  ACRO Biosystems
Mouse MARCO, His Tag MAR-M5245 is expressed from human 293 cells HEK293. It contains AA Gln 70 - Ser 518 Accession Q60754-1. 1 * 100 µG
Catalog # ACRBMAR-M5245100UG
Supplier:  ACRO Biosystems
Frequently Bought Together