ContactUsLinkComponent
SARS-CoV-2 S1 protein N501Y, His Tag S1N-C52Hg is expressed from human 293 cells HEK293. It contains AA Val 16 - Arg 685 Accession QHD43416.1 N501Y. N501Y mutaion was identified in both the SARS-CoV-2 variant known as 20B501Y.V1, VOC 20201201, or B.1 1 * 1 mg
Catalog # ACRBS1N-C52HG-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 S1 protein N501Y, His Tag S1N-C52Hg is expressed from human 293 cells HEK293. It contains AA Val 16 - Arg 685 Accession QHD43416.1 N501Y. N501Y mutaion was identified in both the SARS-CoV-2 variant known as 20B501Y.V1, VOC 20201201, or B.1 1 * 1 mg
Catalog # ACRBS1N-C52HG-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together