ContactUsLinkComponent
Biotinylated Human PSGL-1, His,Avitag PS1-H82E4 is expressed from human 293 cells HEK293. It contains AA Gln 42 - Cys 320 Accession Q14242-1. 1 * 200 µG
Catalog # ACRBPS1-H82E4200UG
Supplier:  ACRO Biosystems
Biotinylated Human PSGL-1, His,Avitag PS1-H82E4 is expressed from human 293 cells HEK293. It contains AA Gln 42 - Cys 320 Accession Q14242-1. 1 * 200 µG
Catalog # ACRBPS1-H82E4200UG
Supplier:  ACRO Biosystems
Frequently Bought Together