Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Mouse IgG2b Fc, Tag Free IGB-M5203 is expressed from human 293 cells HEK293. It contains AA Glu 97 - Lys 335 Accession P01867-2. 1 * 1 mg
Catalog # ACRBIGB-M5203-1MG
Supplier:
ACRO Biosystems
Expand image
Mouse IgG2b Fc, Tag Free IGB-M5203 is expressed from human 293 cells HEK293. It contains AA Glu 97 - Lys 335 Accession P01867-2. 1 * 1 mg
Catalog # ACRBIGB-M5203-1MG
Supplier:
ACRO Biosystems
Frequently Bought Together
Insulin syringes, Omnican® 20
Supplier: B. Braun
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection