ContactUsLinkComponent
SARS-CoV-2 S protein RBD K417T, His Tag SPD-C52Ht is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Accessi on QHD43416.1 K417T. 1 * 1 mg
  ACRBSPD-C52HT-1MG
SARS-CoV-2 S protein RBD K417T, His Tag SPD-C52Ht is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Accessi on QHD43416.1 K417T. 1 * 1 mg
  ACRBSPD-C52HT-1MG
Frequently Bought Together