Atrazine
Glass bottle, 25 g
Catalog # ACRO161680250
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- Bioactive
Synonyms: 2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine
- CAS Number:1912-24-9
- Formula:C₈H₁₄ClN₅
- MDL Number:MFCD00041810
- Molecular Weight:215.69 g/mol
- Melting Pt:175 °C
- Storage Temperature:Ambient
- UN:3077
- ADR:9,III
- :Density: 1.2 g/cm³ (20 °C)
Specifications
Specification Test Results
- Appearance:White to off-white Powder or crystals
- Infrared spectrum:Conforms
- Melting point:173 to 180 °C
- Water:≤1 %
- HPLC:≥95.0 %
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Cat. No.:ACRO161680250



