Atrazine
Glass bottle, 5 g
Catalog # ACRO161680050
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- Bioactive
Synonyms: 2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine
- CAS Number:1912-24-9
- Formula:C₈H₁₄ClN₅
- MDL Number:MFCD00041810
- Molecular Weight:215.69 g/mol
- Melting Pt:175 °C
- Storage Temperature:Ambient
- UN:3077
- ADR:9,III
- :Density: 1.2 g/cm³ (20 °C)
Specifications
Specification Test Results
- Appearance:White to off-white Powder or crystals
- Infrared spectrum:Conforms
- Melting point:173 to 180 °C
- Water:≤1 %
- HPLC:≥95.0 %
Specifications
- Pack type:Glass bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



