ContactUsLinkComponent
CONCENTRATION OF ELEMENT INCLUDES: ELEMENT CONCENTRATION BERYLLIUM 200 µG/ML CERIUM 200 µG/ML IRON 200 µG/ML INDIUM 200 µG/ML LITHIU M 200 µG/ML MAGNESIUM 200 µG/ML LEAD 200 µG/ML URANIUM 200 µG/ML NITRIC ACID 1% 1 * 1 items
Catalog # PERCN8162301
Supplier:  PerkinElmer
CONCENTRATION OF ELEMENT INCLUDES: ELEMENT CONCENTRATION BERYLLIUM 200 µG/ML CERIUM 200 µG/ML IRON 200 µG/ML INDIUM 200 µG/ML LITHIU M 200 µG/ML MAGNESIUM 200 µG/ML LEAD 200 µG/ML URANIUM 200 µG/ML NITRIC ACID 1% 1 * 1 items
Catalog # PERCN8162301
Supplier:  PerkinElmer
Frequently Bought Together