ContactUsLinkComponent
CONCENTRATION OF ELEMENT INCLUDES: ELEMENT CONCENTRATION MOLYBDENUM 3 µG/ML ANTIMONY 1.2 µG/ML TIN 6 µG/ML NITRIC ACID 5 µG/ML HYDRO FLUORIC ACID 1 µG/ML TARTARIC ACID < 1 µG/ML WATER 91.98 µG/ML 1 * 1 items
Catalog # PERCN9304394
Supplier:  PerkinElmer
CONCENTRATION OF ELEMENT INCLUDES: ELEMENT CONCENTRATION MOLYBDENUM 3 µG/ML ANTIMONY 1.2 µG/ML TIN 6 µG/ML NITRIC ACID 5 µG/ML HYDRO FLUORIC ACID 1 µG/ML TARTARIC ACID < 1 µG/ML WATER 91.98 µG/ML 1 * 1 items
Catalog # PERCN9304394
Supplier:  PerkinElmer
Frequently Bought Together