Hypodermic syringes
Hypodermic syringe, with needle, 22G, 10 ml
Supplier: TERUMO
Jump to:
Product Details & Documents
Disposable, with needles.
Dokumente
Specifications
- Capacity:10 ml
- Description:Hypodermic syringe, with needle, 22G
- Seq:07