ContactUsLinkComponent
CHROMABOND columns SiOH BIGpack volume: 6 mL, content of sorbent: 500 mg material: PP, with PE filter elements packed in 5 bags wit h 50 pcs. pack of 250 1 * 250 items
Catalog # MANA730070.250
Supplier:  MACHEREY-NAGEL
CHROMABOND columns SiOH BIGpack volume: 6 mL, content of sorbent: 500 mg material: PP, with PE filter elements packed in 5 bags wit h 50 pcs. pack of 250 1 * 250 items
Catalog # MANA730070.250
Supplier:  MACHEREY-NAGEL