N 20 Aluminium crimp cap, silver, center hole Silicone blue transparent/centrical PTFE-lamination colorless Hardness: 45° shore A T hickness: 3 mm pack of 100 1 * 100 items
Catalog # MANA702340
Supplier: MACHEREY-NAGEL
N 20 Aluminium crimp cap, silver, center hole Silicone blue transparent/centrical PTFE-lamination colorless Hardness: 45° shore A T hickness: 3 mm pack of 100 1 * 100 items
Catalog # MANA702340
Supplier: MACHEREY-NAGEL
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Hydriodic acid 57%, EMSURE® for analysis, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Inoculating loops, reusable
Supplier: RSG
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Erlenmeyer Flasks
Supplier: VWR Collection