Domiphen bromide
Fibre drum, 25 kg
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Domiphen Bromide, BP.
All Spectrum Chemical BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Synonyms: (Dodecyldimethyl-2-phenoxyethyl)ammonium bromide, N,N-Dimethyl-N-(2-phenoxyethyl)dodecan-1-aminium bromide
- CAS Number:538-71-6
- Formula:C₂₂H₄₀BrNO
- MDL Number:MFCD00011769
- Molecular Weight:414.48 g/mol
- Storage Temperature:Ambient
Dokumente
Specifications
Specifications
- Pack type:Fibre drum
- Pk:25 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



