Plastic replacement tips
Plastic replacement tips for forceps, straight, very fine, Stainless steel, 40 mm, Ø 0,5 mm
FSCI11988-02
Marketsource
Pack of (2) PVDF tips and (3) screws. Tips are made of polyvinylidene fluoride (PVDF) and are electrostatic discharge (ESD) safe.
Compatible with Item No. 11980-13.
- Description:Plastic replacement tips for forceps, straight, very fine
- Length:40 mm
- Material:Stainless steel
- Seq:02
- Tip size:Ø 0,5 mm



