Domiphen bromide
100 g
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Domiphen Bromide, BP.
All Spectrum Chemical BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Synonyms: (Dodecyldimethyl-2-phenoxyethyl)ammonium bromide, N,N-Dimethyl-N-(2-phenoxyethyl)dodecan-1-aminium bromide
- CAS Number:538-71-6
- Formula:C₂₂H₄₀BrNO
- MDL Number:MFCD00011769
- Molecular Weight:414.48 g/mol
- Storage Temperature:Ambient
Documents
Specifications
Specifications
- Pk:100 g



