ContactUsLinkComponent
SpectraMax® PARADIGM® Tunable (TUNE) Detection Cartridge is optimized for 96-, 384-, and 1536-well plates and provides wavelength se lection in 1nm increments in Fluorescence Intensity (FI),Time Resolved Fluorescence (TRF), and Luminescence (LUM) de 1 * 1 items
Catalog # MLDV0200-7050
Supplier:  Molecular Devices
SpectraMax® PARADIGM® Tunable (TUNE) Detection Cartridge is optimized for 96-, 384-, and 1536-well plates and provides wavelength se lection in 1nm increments in Fluorescence Intensity (FI),Time Resolved Fluorescence (TRF), and Luminescence (LUM) de 1 * 1 items
Catalog # MLDV0200-7050
Supplier:  Molecular Devices