Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Column 30 Meter x 0.32 mm ID OV-1 Bonded 1.50 um 1 * 1 items
Catalog # HISM130-3210
Supplier:
HICHROM
Expand image
Column 30 Meter x 0.32 mm ID OV-1 Bonded 1.50 um 1 * 1 items
Catalog # HISM130-3210
Supplier:
HICHROM