Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Vivaspin Turbo 15 Regenerated Cellulose membrane, 100,000 MWCO, 12/PK ultrafiltration centrifugal concentrators for protein, DNA, na noparticle and virus concentration for 4-15 mL sample volumes 1 * 12 items
512-0759
:
Sartorius
Vivaspin Turbo 15 Regenerated Cellulose membrane, 100,000 MWCO, 12/PK ultrafiltration centrifugal concentrators for protein, DNA, na noparticle and virus concentration for 4-15 mL sample volumes 1 * 12 items
512-0759
:
Sartorius