FPRM4D FLASH POINT REFERENCE MATERIALS 1 * 200 mL
Catalog # CANN9727-A35
Supplier: CANNON INSTRUMENT
FPRM4D FLASH POINT REFERENCE MATERIALS 1 * 200 mL
Catalog # CANN9727-A35
Supplier: CANNON INSTRUMENT
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
UV/Visible standards, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
pH Standard Solutions, IUPAC, Certified
Supplier: Hach