Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Clear inserts, polyester (PET) membrane, Transwell®

24-well insert plates, 0,33 cm², Polyester, 6,5 mm, 0.4 µm
Catalog # CORN3467
   Low Manufacturing Impact
Supplier:   Corning
Clear inserts, polyester (PET) membrane, Transwell®
Catalog # CORN3467
Supplier:  Corning
   Low Manufacturing Impact
Clear inserts, polyester (PET) membrane, Transwell®
Clear inserts, polyester (PET) membrane, Transwell®

24-well insert plates, 0,33 cm², Polyester, 6,5 mm, 0.4 µm

Price
827,00 €
Log in for availability
1 * 96 ST
Product Details & Documents

Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.

  • w Produce a cell culture environment that closely resembles an in vivo state
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 µm transparent membrane
  • Treated for optimal cell attachment
  • Excellent visibility under phase contrast microscopy
  • Supplied in multiple well plates
  • Sterile

Allow polarised cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally.

Specifications
  • Membrane:
    Polyester
  • Pore size:
    0.4 µm
  • Description:
    24-well insert plates
  • Seq:
    08
  • Culture area:
    0,33 cm²
  • Membrane Ø:
    6,5 mm
  • Cat. No.:
    CORN3467
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Environmentally Preferable:
    Low Manufacturing Impact