Order Entry
Germany
Orders LinkContactUsLinkComponent
Methimazole, Reference Standards

Methimazole, Reference Standards

Methimazole Related Compound C, USP reference standard, 3 x 50 mg, Ampule, 150 mg
Catalog # 1411038.
Supplier:   USP
Methimazole, Reference Standards
Catalog # 1411038.
Supplier:  USP
CAS Number:  
Methimazole, Reference Standards

Methimazole Related Compound C, USP reference standard, 3 x 50 mg, Ampule, 150 mg

CAS Number:  
Price
1 260,00 €
Log in for availability
1 * 150 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Ampule
  • Pk:
    150 mg
  • Standards:
    USP reference standard
  • Description:
    Methimazole Related Compound C
  • Pk Info:
    3 x 50 mg
  • Cat. No.:
    1411038.
  • Description:
    Methimazole Related Compound C
  • Synonym(s):
    1-Methyl-2-(methylthio)-1H-imidazole
  • CAS No.:
    14486-52-3

Specifications