Order Entry
Germany
Orders LinkContactUsLinkComponent
PTFE tapes, ResTape

PTFE tapes, ResTape

ResTape, compatible with gaseous or liquid oxygen, and with many other gases and liquids, Green
Catalog # REST22485
Supplier:   Restek
PTFE tapes, ResTape
Catalog # REST22485
Supplier:  Restek
PTFE tapes, ResTape
PTFE tapes, ResTape

ResTape, compatible with gaseous or liquid oxygen, and with many other gases and liquids, Green

Price
13,64 €
Log in for availability
1 * 1 ST
Product Details & Documents

ResTape PTFE tape is for use with threaded connections in a wide range of plumbing materials.

  • Each roll is 12,7 mm × 6,6 m

Tapes have a maximum effective temperature of 260 °C.

Specifications
  • Colour:
    Green
  • Description:
    ResTape, compatible with gaseous or liquid oxygen, and with many other gases and liquids
  • Seq:
    01