ContactUsLinkComponent
Septum seal silicone/aluminium laminated for GL14 screw caps (hole or closed caps) Ø12.5mm, silicone white/aluminium, 50 shore A, 1. 0m
Catalog # LBCL120-0085892
Supplier:  LABC LABORTECHNIK
Septum seal silicone/aluminium laminated for GL14 screw caps (hole or closed caps) Ø12.5mm, silicone white/aluminium, 50 shore A, 1. 0m
Catalog # LBCL120-0085892
Supplier:  LABC LABORTECHNIK