ContactUsLinkComponent
LF 298 NTC Monitor for direct connection of analogue conductivity measuring cells, with 2 analog and relay outputs, Modbus, 230V and NTC 1 * 1 items
Catalog # WTWO391230
Supplier:  WTW ONLINE
LF 298 NTC Monitor for direct connection of analogue conductivity measuring cells, with 2 analog and relay outputs, Modbus, 230V and NTC 1 * 1 items
Catalog # WTWO391230
Supplier:  WTW ONLINE