ContactUsLinkComponent
Human IL-17 RE (155-454), Fc Tag (ILE-H5256) is expressed from human 293 cells (HEK293). It contains AA Thr 155 - His 454 (Accession Q8NFR9-1). 1 * 100 µG
Catalog # ACRBILE-H5256100UG
Supplier:  ACRO Biosystems
Human IL-17 RE (155-454), Fc Tag (ILE-H5256) is expressed from human 293 cells (HEK293). It contains AA Thr 155 - His 454 (Accession Q8NFR9-1). 1 * 100 µG
Catalog # ACRBILE-H5256100UG
Supplier:  ACRO Biosystems
Frequently Bought Together