Auftragserfassung
Germany
ContactUsLinkComponent
Recombinat Protein,Human Leptin R CD295 Protein, Fc Tag,Human Leptin R, Fc Tag (LER-H5252) is expressed from human 293 cells (HEK29 3). It contains AA Phe 22 - Asp 839 (Accession NP002294.2).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 250 µG
Catalog # ACRBLER-H5252250UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Leptin R CD295 Protein, Fc Tag,Human Leptin R, Fc Tag (LER-H5252) is expressed from human 293 cells (HEK29 3). It contains AA Phe 22 - Asp 839 (Accession NP002294.2).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 250 µG
Catalog # ACRBLER-H5252250UG
Supplier:  ACRO Biosystems
Frequently Bought Together