ContactUsLinkComponent
Biotinylated Human Siglec-10, Fc,Avitag (SI0-H82F6) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Acc ession Q96LC7-1). 1 * 200 µG
Catalog # ACRBSI0-H82F6200UG
Supplier:  ACRO Biosystems
Biotinylated Human Siglec-10, Fc,Avitag (SI0-H82F6) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Thr 546 (Acc ession Q96LC7-1). 1 * 200 µG
Catalog # ACRBSI0-H82F6200UG
Supplier:  ACRO Biosystems
Frequently Bought Together