ContactUsLinkComponent
Human IGF-I R, His Tag (IGR-H5229) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 932 (Accession P08069-1). 1 * 100 µG
Catalog # ACRBIGR-H5229100UG
Supplier:  ACRO Biosystems
Human IGF-I R, His Tag (IGR-H5229) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 932 (Accession P08069-1). 1 * 100 µG
Catalog # ACRBIGR-H5229100UG
Supplier:  ACRO Biosystems
Frequently Bought Together