ContactUsLinkComponent
Recombinat Protein,Human CD55 DAF Protein, His Tag,Human CD55, His Tag (CD5-H5225) is expressed from human 293 cells (HEK293). It co ntains AA Asp 35 - Ser 353 (Accession NP000565.1).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 100 µG
Catalog # ACRBCD5-H5225100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human CD55 DAF Protein, His Tag,Human CD55, His Tag (CD5-H5225) is expressed from human 293 cells (HEK293). It co ntains AA Asp 35 - Ser 353 (Accession NP000565.1).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 100 µG
Catalog # ACRBCD5-H5225100UG
Supplier:  ACROBIOSYSTEMS