ContactUsLinkComponent
Recombinat Protein,Human CD69 CLEC2C Protein, His Tag,Human CD69, His Tag (CD9-H5222) is expressed from human 293 cells (HEK293). It contains AA Ser 62 - Lys 199 (Accession AAH07037).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg
Catalog # ACRBCD9-H5222-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CD69 CLEC2C Protein, His Tag,Human CD69, His Tag (CD9-H5222) is expressed from human 293 cells (HEK293). It contains AA Ser 62 - Lys 199 (Accession AAH07037).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg
Catalog # ACRBCD9-H5222-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together