ContactUsLinkComponent
Recombinat Protein,Human CD72 Protein, Fc Tag,Human CD72, Fc Tag (CD2-H5251) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession AAH30227).This protein carries a human IgG1 Fc tag at the C-terminus. The protein ha 1 * 1 mg
Catalog # ACRBCD2-H5251-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CD72 Protein, Fc Tag,Human CD72, Fc Tag (CD2-H5251) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession AAH30227).This protein carries a human IgG1 Fc tag at the C-terminus. The protein ha 1 * 1 mg
Catalog # ACRBCD2-H5251-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together