ContactUsLinkComponent
Recombinat Protein,Human Contactin-2 Tag1 CNTN2 Protein, His Tag,Human Contactin-2, His Tag (CN2-H5226) is expressed from human 293 cells (HEK293). It contains AA Ser 31 - Asn 1012 (Accession NP005067.1).This protein carries a polyhistidine tag at th 1 * 1 mg
Catalog # ACRBCN2-H5226-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Contactin-2 Tag1 CNTN2 Protein, His Tag,Human Contactin-2, His Tag (CN2-H5226) is expressed from human 293 cells (HEK293). It contains AA Ser 31 - Asn 1012 (Accession NP005067.1).This protein carries a polyhistidine tag at th 1 * 1 mg
Catalog # ACRBCN2-H5226-1MG
Supplier:  ACROBIOSYSTEMS