ContactUsLinkComponent
Recombinat Protein,Human Dkk-1 Protein, Fc Tag,Human Dkk-1, Fc Tag (DK1-H5258) is expressed from human 293 cells (HEK293). It contai ns AA Thr 32 - His 266 (Accession NP036374.1).This protein carries a human IgG1 Fc tag at the C-terminus. The protein 1 * 100 µG
Catalog # ACRBDK1-H5258100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Dkk-1 Protein, Fc Tag,Human Dkk-1, Fc Tag (DK1-H5258) is expressed from human 293 cells (HEK293). It contai ns AA Thr 32 - His 266 (Accession NP036374.1).This protein carries a human IgG1 Fc tag at the C-terminus. The protein 1 * 100 µG
Catalog # ACRBDK1-H5258100UG
Supplier:  ACROBIOSYSTEMS